Learn More
Apexbio Technology LLC FG2216 223387-75-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B58515.1
Encompass_Preferred
FG2216 (CAS 223387-75-5) is a potent inhibitor of hypoxia-inducible factor (HIF) prolyl hydroxylase specifically targeting PDH2 with an IC50 of 3 9 M By inhibiting HIF prolyl hydroxylase FG2216 stabilizes HIF promoting transcriptional activation of genes involved in adaptive responses to hypoxia including endogenous erythropoietin (EPO) production In non-human primate studies oral administration of FG2216 resulted in a marked reversible elevation of circulating EPO levels and stimulated erythropoiesis with a modest increase in fetal hemoglobin FG2216 serves as a valuable research tool for investigating HIF regulation oxygen-sensing pathways and erythropoiesis mechanisms
Catalog No. 50-000-93041
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.