Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-9 Antibody (36912) [FITC], R&D Systems™
SDP

Catalog No. FAB273F
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB273F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB273F Supplier R&D Systems Supplier No. FAB273F

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

FGF-9 Monoclonal antibody specifically detects FGF-9 in Human samples. It is validated for ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)

Specifications

Antigen FGF-9
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 36912
Conjugate FITC
Formulation PBS
Gene Alias FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF-9, Met1-Ser208, Accession # P31371
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2254
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.