Learn More
Enzo Life Sciences FGF-acidic (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2350100
Encompass_Preferred
Alternative name: Fibroblast growth factor 1, FGF-1, HBGF-1, beta-endothelial growth factor, ECCF-beta. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, 150 mM sodium sulfate, pH 7.5. Source: Produced in E. coli. Contains 141 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4036