Learn More
Enzo Life Sciences FGF-acidic (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2350050
Encompass_Preferred
Alternative name: Fibroblast growth factor 1, FGF-1, HBGF-1, beta-endothelial growth factor, ECCF-beta. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, 150 mM sodium sulfate, pH 7.5. Source: Produced in E. coli. Contains 141 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4035
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.