Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z029211
Encompass_Preferred
Fibroblast Growth Factor- acidic (FGF-acidic) also known as FGF-1 and endothelial cell growth factor is a member of the FGF family which currently contain 23 members. FGF acidic and basic unlike the other members of the family lack signal peptides and are apparently secreted by mechanisms other than the classical protein secretion pathway. FGF acidic has been detected in large amounts in the brain. Other cells known to express FGF acidic include hepatocytes vascular smooth muscle cells CNS neurons skeletal muscle cells fibroblasts keratinocytes endothelial cells intestinal columnar epithelium cells and pituitary basophils and acidophils. As with other FGF s FGF acidic exhibits considerable species cross reactivity. FGF acidic and FGF basic stimulate the proliferation of all cells of mesodermal origin and many cells of neuroectodermal ectodermal and endodermal origin.
Catalog No. 50-252-9956
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.