Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0319350
Encompass_Preferred
Fibroblast Growth Factor-basic (FGF-basic) also known as FGF-2 is a pleiotropic cytokine and one of the prototypic members of the heparin-binding FGF family. Like other FGF family members FGF-basic has theBeta trefoil structure. In vivo FGF-basic is produced by a variety of cells including cardiomycotes fibroblasts and vascular cells. FGF-basic regulates a variety of processes including cell proliferation differentiation survival adhesion motility apoptosis limb formation and wound healing. FGF-basic can be tumorigenic due to its role in angiogenesis and blood vessel remodeling. The angiogenic effects of FGF-basic can produce beneficial cardioprotection during acute heart injury.
Catalog No. 50-252-9731
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.