Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547870
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547870 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547870 Supplier Invitrogen™ Supplier No. PA547870
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FGF-19 is observed and less than 5% cross-reactivity with rhFGF-22 and rhFGF-23 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF15
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Fgf15
Gene Accession No. O35622
Gene Alias FGF; Fgf15; FGF-15; FGF19; FGF-19; Fibroblast growth factor; Fibroblast growth factor 15
Gene Symbols Fgf15
Host Species Sheep
Immunogen E. coli-derived recombinant mouse FGF-15 Ser24-Lys218.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14170
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.