Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGF21 Monoclonal Antibody (CL6491)
GREENER_CHOICE

Catalog No. PIMA531435 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531435 Supplier Invitrogen™ Supplier No. MA531435
  • $630.00
    $510.65 / Each of 1

    Save $119.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Immunogen sequence: PEPPGILAPQ PPDVGSSDPL SMVGPSQGR Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 71%.

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF21
Applications Western Blot
Classification Monoclonal
Clone CL6491
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGF21
Gene Accession No. Q9NSA1
Gene Alias FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Gene Symbols FGF21
Host Species Mouse
Immunogen Synthetic peptide corresponding to Human FGF21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.