Learn More
FGF21 Mouse anti-Human, Clone: OTI3G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502785

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Specifications
FGF21 | |
Monoclonal | |
Unconjugated | |
FGF21 | |
fibroblast growth factor 21 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26291 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI3G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NSA1 | |
FGF21 | |
Human recombit protein fragment corresponding to amino acids 29-209 of human FGF21 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.