Learn More
Invitrogen™ FGF9 Monoclonal Antibody (FG9-77)
Description
Recommended positive controls: recombinant human FGF-9.
Specifications
Specifications
| Antigen | FGF9 |
| Applications | Dot Blot, ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | FG9-77 |
| Concentration | 2.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FGF9 |
| Gene Accession No. | P31371, P54130 |
| Gene Alias | Eks; elbow knee synostosis; FGF; Fgf9; FGF-9; Fibroblast growth factor; Fibroblast growth factor 9; fibroblast growth factor 9 (glia-activating factor); GAF; glia activating factor; glia-activating factor; HBFG-9; HBGF-9; Heparin-binding growth factor 9; M-FGF-9; SYNS3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.