Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGFR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA131990
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA131990 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA131990 Supplier Invitrogen™ Supplier No. PA131990
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Jurkat cell lysate Store as concentrated solution. Centrifuge briefly prior to opening vial.

FGFR2 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR family members differ from one another in their ligand affinities and tissue distribution. Mutations in this gene are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Beare-Stevenson cutis gyrata syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. Multiple alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene FGFR2
Gene Accession No. P21802, P21803
Gene Alias AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor; FGF Receptor 2; FGF-10; FGFR; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25
Gene Symbols FGFR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 22-51 amino acids from the N-terminal region of human FGFR2.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14183, 2263
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.