Learn More
ACROBiosystems Recombinant Protein;Mouse FGF R4 / CD334 Protein;HEK293;1MG;FG4,FGFR4,FGFR-4,CD334,JTK2,MGC20292,TKF
Supplier:  ACROBiosystems FG4M52HA1MG
Encompass_Preferred
Recombinant Protein;Mouse FGF R4 / CD334 Protein;1MG;Mouse FGF R4 (FG4-M52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 17 - Asp 366 (Accession # Q03142-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.7 kDa. The protein migrates as 55-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse FGF R4 (Cat. No. FG4-M52Ha) at 2 μg/mL (100 μL/well) can bind Human FGF-9, Fc Tag (Cat. No. FG9-H4267) with a linear range of 0.156-5 μg/mL (QC tested).
Catalog No. 50-165-7844
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.