Learn More
AdipoGen FGL1 (human) ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45B0022KI01
Encompass_Preferred
ELISA Kit. Assay Type: Sandwich. Detection Type: Colorimetric. Sample Type: Cell Culture Supernatant,Plasma,Serum. Range: 7.8 to 500pg/ml. Sensitivity: 1.8pg/ml. FGL1 has been shown to be upregulated in human cancers and FGL1 is a major functional ligand of LAG-3. FGL1 interacts with LAG-3 in an MHC-II-independent manner and this interaction involves the FGL1 fibrinogen-like domain and the LAG-3 D1-D2 domain. FGL1-LAG-3 interaction blockade promotes tumor immunity by stimulating T cell expansion and activation. FGL1 forms two disulfide-linked homodimers and also higher molecular weight homooligomers that bind to LAG-3 much better than the dimeric forms. This binding to LAG-3 inhibits T cell responses. FGL1 is normally released by the liver in low levels but at higher levels in certain cancer such as non-small cell lung carcinomas (NSCLC) . Upregulated FGL1 in human cancers is associated with a poor prognosis.
Catalog No. 50-205-3329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.