Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FH 535, Tocris Bioscience™

Catalog No. 434410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-441-0 10 mg
43-445-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-441-0 Supplier R&D Systems Supplier No. 4344/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Inhibitor of Wnt/β-catenin signaling

Inhibitor of Wnt/β-catenin signaling and dual antagonist of PPARγ/δ activity. Suppresses β-catenin/Tcf-mediated transcription; inhibits β-catenin and GRIP1 recruitment to PPARγ and δ. Exhibits antiproliferative effects in transformed colon, lung and liver cancer cell lines.

Chemical Identifiers

CAS 108409-83-2
Molecular Formula C13H10Cl2N2O4S
Molecular Weight (g/mol) 361.19
MDL Number MFCD01212888
InChI Key AXNUEXXEQGQWPA-UHFFFAOYSA-N
Synonym 2,5-dichloro-n-2-methyl-4-nitrophenyl benzenesulfonamide, beta-catenin/tcf inhibitor, fh535, n-2-methyl-4-nitrophenyl-2,5-dichlorobenzenesulfonamide, cbmicro_011295, fh hplc
PubChem CID 3463933
IUPAC Name 2,5-dichloro-N-(2-methyl-4-nitrophenyl)benzene-1-sulfonamide
SMILES CC1=C(NS(=O)(=O)C2=C(Cl)C=CC(Cl)=C2)C=CC(=C1)[N+]([O-])=O

Specifications

Quantity 10 mg
Formula Weight 361.2
Percent Purity >99%
Chemical Name or Material FH 535
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.