Learn More
Apexbio Technology LLC FH535, 50mg. Cas: 108409-83-2 MFCD: MFCD01212888. Wnt/B-catenin inhibitor
SDP
Supplier:  Apexbio Technology LLC A341350
Encompass_Preferred
FH535 is a small molecule inhibitor of Wnt/B-catenin with IC50 values of 15.4M, 10.9M, 9.3M, respectively in LCSC, Huh7, PLC cell lines [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4801
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.