Learn More
Thermo Scientific™ FHZ 672 E10 NBR Detector
NBR Detector for the sensitive detection of artificial sources in fluctuating natural radiation fields. Includes black aluminum housing with handle and holder for FH40.
Supplier: Thermo Scientific™ 425406650
This item is not returnable.
View return policy
Description
Specifications
| 48 keV | |
| Aluminum | |
| NBR Detector |