Learn More
Medchemexpress LLC Fialuridine | 69123-98-4 | MFCD00866922 | 99.9% | 372.09 g/mol | C9H10FIN2O5 | 5 MG
Supplier: Medchemexpress LLC HY1181225MG
Fialuridine (FIAU) is a nucleoside analog research compound with reported activity against hepatitis B virus, orthopoxviruses, and herpes simplex viruses. It is supplied as a milligram-scale solid for laboratory research and is not for clinical use.
- Nucleoside analog with reported antiviral activity against HBV, orthopoxviruses, and HSV-1/HSV-2.
- High purity: 99.91% (as provided by the supplier).
- Molecular formula C9H10FIN2O5 and molecular weight ~372.09 g/mol.
- Supplied as a 5 mg solid suitable for small-scale experiments.
- Comes with a supplier SDS containing handling and safety information.