Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIBB Recombinant Mouse Monoclonal Antibody (8E5)
GREENER_CHOICE

Catalog No. PIMA548077
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548077 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548077 Supplier Invitrogen™ Supplier No. MA548077
Only null left
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: 8E5 binds to the amino-terminal region of the Beta-chain of human fibrin at an epitope consiting of GHRPLDKC. Binding is possible in both the monomeric form and the polymeric form of the protein. The epitope recognised by this clone is only accessible upon conversion of fibrinogen to fibrin, which entails the proteolytic removal of fibrinopeptides A & B from fibrinogen by thrombin. The antibody is there is therefore fibrin specific. Fibrin is a non-globular protein which plays a role in the clotting of blood and circulates as the precursor fibrinogen. Polymerization occurs through the action of the protease thrombin, which results in the active fibrin. Fibrin polymerizes as an insoluble matrix and forms a clot along with platelets over a wound site.

FIBB is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in its gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIBB
Applications ELISA, Functional Assay, Immunomicroscopy
Classification Recombinant Monoclonal
Clone 8E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Fgb
Gene Accession No. P02675
Gene Alias 2510049G14Rik; Ab1-181; Ab1-216; Ac1-581; B beta polypeptide; beta-fibrinogen; epididymis secretory sperm binding protein Li 78p; Fgb; fibrinogen; Fibrinogen beta chain; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; Fibrinopeptide B; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC104327; MGC120405
Gene Symbols Fgb
Host Species Mouse
Immunogen 8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.