Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers

Catalog No. 13994104 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
100 (10 x 10 Grid)
16 (4 x 4 Grid)
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Quantity
13-994-104 100 (10 x 10 Grid) Each
11-678-24C 100 (10 x 10 Grid) Each
11-678-26D 49 (7 x 7 Grid) 12 Pack
13-992-33 64 (8 x 8 Grid) 12 Pack
13-989-218 81 (9 x 9 Grid) 12 Pack
13-994-081 16 (4 x 4 Grid) Each
13-994-085 25 (5 x 5 Grid) Each
13-994-091 49 (7 x 7 Grid) Each
13-994-092 64 (8 x 8 Grid) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-994-104 Supplier Thermo Scientific™ Supplier No. 820100
  • $963.00
    $765.00 / Case of 100

    Save $198.00

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organize vial storage within cryo boxes with these fiberboard box dividers.

Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.

  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
Holds 100 (10 x 10 Grid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.