Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fiberlite™ F14-6 x 250y Fixed Angle Rotor
GSA_VAGREENER_CHOICE

Catalog No. p-4528135
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
78-500 Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 78-500 Supplier Thermo Scientific™ Supplier No. 78500
Only null left
Add to Cart
Add to Cart
Request a Quote

Process large volumes with the six-place, 250 mL fixed angle rotor.

Process large volumes with the six-place, 250mL Thermo Scientific™ Fiberlite™ F14-6x250y Fixed Angle Rotor.

Features:

Fiberlite rotors:

  • Run times are reduced with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion- and fatigue-resistant design ensures structural integrity
  • Rotor is backed by a 15-year warranty

Compatible with:

Thermo Scientific™ Sorvall™ RC-6 Plus, Sorvall Evolution™ RC Series, Thermo Scientific™ Jouan KR22i, Beckman™ J2 Series, Beckman J2, Avanti Series, Hitachi™ Himac CR21/CR22 Series, Nalgene™ bottles and tubes

Recommended for:

Processing lysates, homogenates, ammonium sulphate precipitation of proteins and enzymes and plant tissue homogenates

†Performance may vary by centrifuge model.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Fiberlite F14-6 x 250y Fixed Angle Rotor
Material Carbon Fiber Composite
Type Fixed Angle Rotor
Capacity 6 x 250 mL
Max. RCF 30,100 x g
Max. Speed 14,000 rpm
Radius (Metric) 137 mm
Radius (Metric) Min. 37 mm
K Factor 1690
Max. Tube Length 135 mm
Max. Tube Diameter 62 mm
Net Weight 15.5 lb. (7.03 kg)
For Use With (Equipment) Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges
Angle 23°
Min. RCF 8,122 x g
Biocontainment Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.