Learn More
Honeywell™ Fibre-Metal™ Roughneck™ Hard Hat
Catalog No. 19811520 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Blue
Green
White
Suspension Type:
8-point 3R Rachet
8-point SwingStrap™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-811-520 Blue 8-point 3R Rachet
19-811-517 Green 8-point 3R Rachet
19-811-521 White 8-point SwingStrap™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-811-520 Supplier Honeywell™ Supplier No. P2AQRW71A000
Add to Cart
Add to Cart
Specifications
Certifications/Compliance ANSI Z89.1-2009 Type 1 Class G and CSA Z94.1-2005 Type 1 Class C
Construction Injection-molded fiberglass
Color Blue
Finish Smooth Matte
Includes Quick-Lok™ Connection
Material Fiberglass
Quantity 1 Ea.
Style Roughneck
Suspension Type 8-point 3R Rachet
Weight (English) 1 lb.
Weight (Metric) 0.5 kg