Learn More
Honeywell™ Fibre-Metal™ Roughneck™ Hard Hat
Catalog No. 19811521 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Blue
Green
White
Suspension Type:
8-point 3R Rachet
8-point SwingStrap™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-811-521 White 8-point SwingStrap™
19-811-520 Blue 8-point 3R Rachet
19-811-517 Green 8-point 3R Rachet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-811-521 Supplier Honeywell™ Supplier No. P2AQSW01A000
  • $129.00
    $86.65 / Each of 1
    Save $42.35
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Specifications
Certifications/Compliance ANSI Z89.1-2009 Type 1 Class G and CSA Z94.1-2005 Type 1 Class C
Construction Injection-molded fiberglass
Color White
Finish Smooth Matte
Includes Quick-Lok™ Connection
Material Fiberglass
Quantity 1 Ea.
Style Roughneck
Suspension Type 8-point SwingStrap™
Weight (English) 1 lb.
Weight (Metric) 0.5 kg