Learn More
Honeywell Fibre-Metal™ Fibre-Metal™ Roughneck P2 Fiberglass Cap Style Hard Hat With SuperEight™ Rachet
Catalog No. 19085625 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Width (Metric):
22.86 cm
Catalog No. Width (Metric)
19-085-625 22.86 cm
1 options
Catalog No. 19-085-625 Supplier Honeywell Fibre-Metal™ Supplier No. P2HNQRW09A000
Add to Cart
Add to Cart
The industry standard for heavy duty protection, rated for temperatures up to 500°C. Comes with attached Quick Lok blocks for ease in attaching faceshields or welding helmets.
  • Exceptional impact and crack resistance surpassing many performance tests
  • The Roughneck P2A proprietary injection-molded fiberglass compound improves durability and is eight times stronger than common high density polyethylene (HDPE) cap shells
  • Improved heat resistance meets ANSI-Z89.1-2009 impact resistance tests at extreme temperatures of up to 500°F
  • Injection-molded solid color resin provides a permanent, no-chip, no-peel finish
  • Smooth, matte finish makes logo imprints crisper and cleaner to make official company caps look better and last longer
  • Built for strength, toughness and durability
  • Available in nine popular colors
Specifications
Certifications/Compliance ANSI Z89.1-2009 Type I Class G, CSA Z94.1-2005 Type I Class G or C
Color Gray
Material Fiberglass
Product Type Hard Hat
Size One Size Fits Most
Suspension Type 8-Point Ratchet Suspension
Ventilated Not Ventilated
Head Style Short Brim (Cap Style)
Width (Metric) 22.86 cm