Learn More
Honeywell™ Fibre-Metal™ Supereight™ 3-R Ratchet Headband
Catalog No. 19017843 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Blue
Gray
Hi-vis Orange
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-017843 Yellow
19-017837 Gray
19-017840 Hi-vis Orange
19-017836 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-017843 Supplier Honeywell™ Supplier No. E2RW02A000
  • $62.75
    $42.65 / Each of 1
    Save $20.10
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Specifications
Certifications/Compliance ANSI-Z89.1 TYPE 1, Class E, G, or C
Color Yellow
Finish Smooth
Material Superlectric Thermoplastic
Quantity 1 Ea.
Suspension Type 8-point 3R Rachet
Head Style Smooth Crown