Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Fibre-Metal™ Visors for F400 Headgear

Catalog No. 19811502 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Height (English):
8 in.
8.75 in.
Lens Tint:
Clear
Shade 5
Width (English):
16.5 in.
19 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Lens Tint Width (English)
19-811-502 8 in. Shade 5 16.5 in.
19-017927 8 in. Clear 16.5 in.
19-017929 8.75 in. Clear 19 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-811-502 Supplier Honeywell™ Supplier No. 4178IRUV5
  • $51.10
    $34.65 / Each of 1

    Save $16.45

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Height (English) 8 in.
Width (English) 16.5 in.
Lens Tint Shade 5
For Use With (Equipment) F400 headgear in.
Lens Material Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.