Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrillin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552995 Supplier Invitrogen™ Supplier No. PA552995
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMCFSGLVNG RCAQELPGRM TKMQCCCEPG RCWGIGTIPE ACPVRGSEEY RRLCMDGLPM GGIPGSAGSR PGGTGGNGFA PSGNGNGYGP GGTGFIPIPG GNGFSPGVGG AGVGAGGQGP IITGLTILNQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

The protein encoded by this gene is a component of connective tissue microfibrils and may be involved in elastic fiber assembly. Mutations in this gene cause congenital contractural arachnodactyly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillin 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBN2
Gene Accession No. P35556
Gene Alias BC063774; CCA; DA9; EOMD; Fbn2; Fbn-2; Fib-2; fibrillin 2; fibrillin 5; fibrillin-2; Fibrillin-2 C-terminal peptide; mKIAA4226; sne; sy; syndatyly ems
Gene Symbols FBN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Fibrillin 2. Recombinant protein control fragment (Product #RP-90288).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.