Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fibrinogen-Binding Inhibitor Peptide

Catalog No. AAJ60863LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60863LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60863LB0 Supplier Thermo Scientific Chemicals Supplier No. J60863LB0
Add to Cart

For cellular and molecular biology applications

Inhibits the binding of fibrinogen and fibronectin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name Fibrinogen γ-chain dodecapeptide
Format Solid
For Use With (Application) Inhibits the binding of fibrinogen and fibronectin
Molecular Weight (g/mol) 1189.30
Product Type Fibrinogen-Binding Inhibitor Peptide
Quantity 5 mg
Regulatory Status RUO - research use only
Sequence H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.