Learn More
CPC Scientific H-Glu-His-Ile-Pro-Ala-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific FBP001B
Encompass
Fibrinogen is a cofactor in platelet activation. This peptide binds to fibrinogen and inhibits the adhesion of platelets to fibrinogen, platelet aggregation and the adhesion of platelets to vitronectin. The peptide has the characteristics expected of a mimic of a glycoprotein IIb/IIIa ligand binding site.SEQUENCE: H-Glu-His-Ile-Pro-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: EHIPAMOLECULAR FORMULA: C25H39N7O8MOLECULAR WEIGHT: 565.63STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [137235-80-4]SYNONYMS: Fibrinogen Binding PeptideRESEARCH AREA: CardiovascularREFERENCES:T.K.Gartner and D.B.Taylor, Proc. Soc. Exp. Biol. Med., 198, 649 (1991)SKU(s): FBP-001A, FBP-001B
Catalog No. 50-210-9272
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.