Learn More
Description
MA5-15906 targets FGG in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15906 immunogen is purified recombinant fragment of human FGG expressed in E. Coli. . MA5-15906 detects FGG which has a predicted molecular weight of approximately 52kDa.
Specifications
Specifications
| Antigen | Fibrinogen gamma |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4H9 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | Fgg |
| Gene Accession No. | P02679, Q8VCM7 |
| Gene Alias | 3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
