Learn More
Innovative Research Inc Human Fibrinogen Purified Biotin Labeled 10mg
Supplier:  Innovative Research Inc IHUFBGBL10MG
Encompass_Preferred
Human Fibrinogen Purified from Innovative Research has been purified from human plasma by a propritetary process that combines multiple conventional chromatographic methods. This is a Frozen liquid buffered in 20mM Sodium Citrate-HCl; pH 7.4 with a concentration of 40 mg/ml and a purity of >95% as determined by SDS-PAGE.
Catalog No. 50-203-6601
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.