Learn More
Diagnostic Biosystems FIBRONECTIN Fibronectin
Supplier:  Diagnostic Biosystems RP01305
Encompass_Preferred
This antibody reacts with human fibronectin in liver, tonsil, skin and kidney. Traces of contaminating antibodies have been removed by solid-phase adsorption. Only the fibronectin arch was observed in crossed immunoelectrophoresis.
Catalog No. 50-238-5712
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.