Learn More
Medchemexpress LLC HY-P0306 10mg Medchemexpress, Fibronectin Adhesion-promoting Peptide CAS:125720-21-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP030610MG
Encompass_Preferred
Medchemexpress, HY-P0306 10mg Fibronectin Adhesion-promoting Peptide CAS:125720-21-0 Fibronectin Adhesion-promoting Peptide (Heparin Binding Peptide) is one of the heparin-binding amino acid sequences found in the carboxy-terminal heparin-binding domain of fibronectin. It promotes assembly of mesenchymal stem cell (MSC) spheroids into larger aggregates. Fibronectin Adhesion-promoting Peptide directly promotes the adhesion, spreading, and migration of endothelial cells by reacting with heparin binding domains of cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3034