Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibulin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA131208
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA131208 100 μg
1 options

Catalog No. PIPA131208

Supplier: Invitrogen™ PA131208

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Human RPE cell extracts..

This gene encodes a member of the fibulin family of extracellular matrix glycoproteins. Like all members of this family, the encoded protein contains tandemly repeated epidermal growth factor-like repeats followed by a C-terminus fibulin-type domain. This gene is upregulated in malignant gliomas and may play a role in the aggressive nature of these tumors. Mutations in this gene are associated with Doyne honeycomb retinal dystrophy. Alternatively spliced transcript variants that encode the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibulin 3
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Tris glycine with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.9
Gene EFEMP1
Gene Accession No. O35568, Q12805, Q8BPB5
Gene Alias DHRD; DRAD; EFEMP1; EGF containing fibulin like extracellular matrix protein 1; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; epidermal growth factor-containing fibulin-like extracellular matrix protein 1; extracellular protein S1-5; FBLN3; FBNL; FIBL-3; Fibrillin-like protein; Fibulin-3; FLJ35535; MGC111353; MLVT; MTLV; S1-5; T16
Gene Symbols EFEMP1
Host Species Rabbit
Immunogen Synthetic cyclic peptide corresponding to N terminal amino acids 25-42 of Human EFEMP1, peptide was covalently modified.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216616, 2202, 305604
Target Species Human, Mouse, Monkey, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.