Description
Includes
Secchi disk with line and float; -40/+120°F armored thermometer; 8 dia. x 20 in.L plankton net; 1.75L water bottle; 125mL polyethylene wash bottles (12); 250mL polypropylene square wash bottles (12); 500mL polyethylene one-piece wash bottle for rinsing net; set of comprehensive teacher instructions
Durable, reusable apparatus.
- Basic water bottle needs no messenger; constructed of PVC with soft rubber seals and 20m of line on a styrofoam float that will bob to the surface if dropped
- Plankton net constructed of genuine Nitex™ 80 micron mesh with stainless steel ring and bridle for attaching line
- Secchi disk determines turbidity in shallow waters and meets industry standards
Specifications
Specifications
| For Use With (Application) | Obtaining samples of phyto and zooplankton from lakes and ponds for later study in the lab |
| Includes | Secchi disk with line and float; -40/+120°F armored thermometer; 8 dia. x 20 in. L plankton net; 1.75 L water bottle; 125 mL polyethylene wash bottles (12); 250 mL polypropylene square was bottles (12); 500 mL polyethylene one-piece wash bottle for rinsing net; set of comprehensive teacher instructions |