Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Filamin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545573 Supplier Invitrogen™ Supplier No. PA545573
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VGKSADFVVE AIGTEVGTLG FSIEGPSQAK IECDDKGDGS CDVRYWPTEP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes one of three related filamin genes, specifically gamma filamin. These filamin proteins crosslink actin filaments into orthogonal networks in cortical cytoplasm and participate in the anchoring of membrane proteins for the actin cytoskeleton. Three functional domains exist in filamin: an N-terminal filamentous actin-binding domain, a C-terminal self-association domain, and a membrane glycoprotein-binding domain. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filamin 2
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FLNC
Gene Accession No. Q14315
Gene Alias 1110055E19Rik; ABP-280; ABP280A; ABP-280-like protein; ABPA; ABPL; ABP-L; ABP-L, gamma filamin; actin binding protein 280; actin-binding protein 280; actin-binding-like protein; filamin C; Filamin C gamma; filamin C, gamma; filamin C, gamma (actin binding protein 280); filamin, gamma; filamin-2; Filamin-C; FLJ10186; FLN2; FLNC; FLN-C; gamma filamin; gamma-filamin; MFM5; MPD4
Gene Symbols FLNC
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FLNC (aa 586-635).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2318
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.