Learn More
Selleck Chemical LLC Filgotinib (GLPG0634) 5mg 1206161-97-8
Supplier: Selleck Chemical LLC S76055MG
Filgotinib (GLPG0634) is a selective JAK1 inhibitor with IC50 of 10 nM, 28 nM, 810 nM, and 116 nM for JAK1, JAK2, JAK3, and TYK2, respectively. Phase 2. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.