Learn More
Cayman Chemical Filipin III, 480-49-9, 500 g
SDP
Supplier:  Cayman Chemical 70440500
Encompass_Preferred
Molecular Formula C35H58O11, Purity ≥90%, Formula Weight 654.8; A fluorescent molecule that can be used to detect cholesterol by fluorescence and freeze-fracture electron microscopy.
Catalog No. NC9860572
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.