Learn More
Apexbio Technology LLC Filipin III 480-49-9 5mg
SDP
Supplier:  Apexbio Technology LLC B60345
Encompass_Preferred
Filipin III is a naturally occurring fluorescent polyene macrolide antibiotic isolated as the primary active isomer from cultures of Streptomyces filipinensis It binds specifically to free (unesterified) cholesterol in biological membranes through reversible interaction causing alteration of its spectral properties (excitation/emission maxima around 360 nm/480 nm) Due to this cholesterol-binding affinity and inherent fluorescence Filipin III has been extensively employed as a probe for visualizing and quantifying cholesterol distribution and trafficking in cell membranes In biomedical research Filipin III is applied in fluorescence microscopy and flow cytometry to detect free cholesterol dynamics lipid raft formation and lipoprotein metabolism alterations Currently no definitive IC50 values have been reported for Filipin III in typical cellular assay conditions
Catalog No. 50-000-63652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.