Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Filled Five-Shelf Cabinet

Catalog No. 19090663 Shop All Medique Products
Change view
Click to view available options
Dimensions (L x W x H):
28.12 x 19.25 x 6.62 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
19-090-663 28.12 x 19.25 x 6.62 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-663 Supplier Medique Supplier No. 738ANSI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For large industrial work settings.

  • Convenient door pouches
  • Extra-large cabinet
  • Contains assortment of adhesive bandages, dressing, eye care products, antiseptics, employee comfort items, etc

Specifications

For Use With (Application) Large Industrial Work Settings
Product Type Filled Cabinet
Includes Assortment Of Adhesive Bandages, Dressing, Eye Care Products, Antiseptics, Employee Comfort Items
Model Five-shelf
Dimensions (L x W x H) 28.12 x 19.25 x 6.62 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.