Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Filling Tube, 190 mm

Catalog No. 01915182 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Densito and DensitoPro Handheld Density Meters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-915-182 Densito and DensitoPro Handheld Density Meters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-915-182 Supplier Mettler Toledo™ Supplier No. 30330847
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mettler Toledo Filling Tube, 190 mm is designed for use with Densito or DensitoPro.

  • For use with Densito and DensitoPro
TRUSTED_SUSTAINABILITY

Specifications

Languages Chinese, English (GB), English, French, German, Italian, Korean, Polish, Brazilian Portuguese, Russian, Spanish
Description Sampling Tube, 190 mm
Height (Metric) 9 cm
For Use With (Equipment) Densito and DensitoPro Handheld Density Meters
For Use With (Application) Use with Densito and DensitoPro Handheld Density Meters
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.