Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Filling Tubes for BRAND™ Bottletop Dispensers and Burettes

Catalog No. 13880729 Shop All BrandTech Scientific Products
Change view
Click to view available options
Length (Metric):
125 to 240 mm
170 to 330 mm
195 to 350 mm
250 mm
250 to 480 mm
500 mm
70 to 140 mm
Sterility:
Non-sterile
Sterile
For Use With (Equipment):
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
Seripettor™ (All Volumes)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Sterility For Use With (Equipment)
13-880-729 500 mm Sterile Seripettor™ (All Volumes)
14-380-911 70 to 140 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14-380-912 125 to 240 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14-380-913 195 to 350 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14-380-914 250 to 480 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
14-380-915 170 to 330 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
14-380-916 250 to 480 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
03-840-95 250 mm Non-sterile Seripettor™ (All Volumes)
03-840-94 500 mm Non-sterile Seripettor™ (All Volumes)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-880-729 Supplier BRANDTECH™ BRAND™ Supplier No. 704538
Add to Cart
Edge
Add to Cart

Replacement filling tubes for BRAND™ bottletop instruments, including Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis dispensers; seripettor™ and seripettor™ pro dispensers; and Titrette™ bottletop burettes.

  • Telescoping FEP filling tubes adjust to bottle height without cutting
    • Compatible with Dispensette™ S, Dispensette™ S Organic, Dispensette™ S Trace Analysis; seripettor™ pro; and Titrette™
    • Also compatible with Dispensette™ III
  • Standard PP filling tubes can be manually cut to fit various bottle sizes
    • Compatible only with seripettor™
    • Available sterile with O-ring
  • Available in multiple lengths based on dispenser nominal volume
  • Design supports aspiration from the bottom of the bottle while reducing precipitate intake
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polypropylene
For Use With (Equipment) Seripettor™ (All Volumes)
Sterility Sterile
Length (Metric) 500 mm
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.