Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Valve Mounting Tool for Bottletop Dispensers

Catalog No. 13880704 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Dispensette S instruments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-880-704 Dispensette S instruments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-880-704 Supplier BRANDTECH™ BRAND™ Supplier No. 6687

missing translation for 'validMainframeMsg'

Valve mounting tool for BRAND Dispensette III and Organic

Calibrating-mounting tool is used for calibrating analog-adjustable Dispensette™ models, as well as removing the valves for routine cleaning or replacement.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Microfilter Connector
Autoclavable Autoclavable
Color Black
Material Polypropylene
For Use With (Application) Adjusting Dispensette S
For Use With (Equipment) Dispensette S instruments
Includes Tool only
Sterility Non-sterile
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.