Learn More
BRANDTECH™ BRAND™ Replacement filling valves for BRAND™ Bottletop Dispensers
Shop All BrandTech Scientific ProductsDescription
- Filling valves for the Dispensette™ III, Organic, TA and seripettor™ pro are provided with a sealing washer. The updated design of the Dispensette™ S series of bottletop dispensers has eliminated the need for a sealing washer.
- Filling valve with olive-shaped nozzle is recommended for Dispensette™ III and Organic bottletop instruments that are frequently autoclaved.
- seripettor™ valve set includes both the filling and discharge valves for the instrument.
- Calibrating-mounting tool is used for calibrating analog-adjustable Dispensette™ models, as well as removing the valves for routine cleaning or replacement.
Specifications
Specifications
| Product Type | Filling Valve |
| Autoclavable | Autoclavable |
| Color | Translucent/White |
| Material | ETFE, Borosilicate Glass, Ceramic |
| For Use With (Equipment) | Dispensette S/S Organic - 25, 50, and 100 mL |
| Includes | Filling Valve |
| Sterility | Non-sterile |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.