Learn More
MTC Bio Blu-Lite HS Autoradiography film, 5x7in, 100 sheets/box
Supplier:  MTC Bio A8813
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Blu-Lite UHC™ is MTC Bios newest line of high contrast, high sensitivity autoradiography film. Blu-Lite UHCfilm is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications. In addition to Blu-Lite UHCs excellent sensitivity and contrast characteristics, it is also coated on both sides (double emulsion) for improved consistency and overall performance.
Catalog No. 50-112-7124