Learn More
Grainger Film Tape 1/4 in x 5 yd Amber 2.5 mil
Supplier:  Grainger 15C588
Encompass_Preferred
The TAPECASE Film Tape 1/4 in x 5 yd is a polypropylene film tape with an acrylic adhesive. It has a thickness of 1.4 mil and a width of 1/4 inch. The tape length is 5 yards and it is transparent in color. This tape is designed for general purpose holding, splicing, sealing and masking applications. It can adhere to paper, cardboard, glass, plastic and metal surfaces. The operating temperature range is up to 200°F.
Catalog No. 50-275-1797