Learn More
Microsolv Technology Corporation Film, Zone Free, Sealing for Micro-Sample Management System or 96 Well Plates. 70um polyethylene top layer and inert white layer / Adhesive Free Zones. 50/CS.
Supplier:  Microsolv Technology Corporation 95296ZFFCS
Encompass_Preferred
Film has 3.12" x 6" x 6" outer dimensions. Supplied in a 4" x 7" bag.
Catalog No. 50-227-0498
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.