Learn More
Agilent BioTek Filter Cube, GFP excitation - CY5 emission
Shop All Agilent Technologies Inc. Products
Description
- Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
- Requires 465 nm LED cube (1225001)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Equipment) | Microplate readers and imagers |