Learn More
DWK Life Sciences Kimble™ Filter Flasks Spare Parts - Plastic Sidearm and Bushing
Shop All DWK Life Sciences Products

Description
- Detachable autoclavable plastic sidearm designed to accept 5/16 inch ID flexible tubing (U.S. Patent 3,268,300)
- Sidearm has two ears to provide a finger grip for easy insertion into a neoprene bushing by turning the plastic piece
- Opening in the sidearm is larger at the flask end to hold a cotton plug securely
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | filtration |
| For Use With (Equipment) | Filter Flasks |
| Includes | Neoprene bushing |
| No. per Pack | 1 |
| Product Type | Plastic Sidearm and Bushing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.