Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Filter Funnel
GREENER_CHOICESDP

Catalog No. 31502383 Shop All Kemtech Products
Change view
Click to view available options
Diameter (Metric)Top:
50 mm
75 mm
90 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top
31-502-383 90 mm
31-502-381 50 mm
31-502-382 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-383 Supplier Synthware™ Supplier No. F932490X
Add to Cart
Add to Cart

Funnel features a fritted disc and can be used to filter particulates from a variety of substances.

  • Filter funnel with a fritted disc
  • Use to filter particulates from a variety of substances

Specifications

Diameter (Metric)Top 90 mm
Material Borosilicate Glass (3.3)
Color Clear
For Use With (Application) Filtering Media
Joint Number 24/40
Porosity Extra Coarse
Product Type Filter Funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.