Learn More
Caron Products And Services PM Kit Model CRSY102 Condensate Recirculator
Supplier:  Caron Products And Services PMCRSY102
Encompass_Preferred
Preventative Maintenance Kit for Model CRSY102 (Condensate Recirculator) (Replaces FLTR101) Includes (1) CLP-130 SS Clip, (1) DEM-103 Demineralizer Cartridge, (1) FIL-607 Carbon Cartridge, (1) HUM-120 Copper Mesh Antimicrobial Rod, (1) NOZ-110 Atomizer, Brass with Mesh Screen, and (1) SIL-111 Silicone Grease. Detailed Instructions Included.
Catalog No. NC9667361
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.